Learn More
Cell Biolabs IncSupplier Diversity Partner PSCAAV-GFP CONTROL PLASMID  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner AAV410
pscAAV-GFP Control Vector may be co-transfected with AAV packaging plasmids to generated recombinant adeno-associated virus.
Catalog No. 50-672-460