Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PSD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542177
Description
Peptide sequence: SDVAKHLGKN NEFSKLVAEE YLKFFDFTGM TLDQSLRYFF KAFSLVGETQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
Guanine nucleotide exchange factor for ARF6.
Specifications
PSD3 | |
Polyclonal | |
Unconjugated | |
PSD3 | |
4931420C21Rik; ADP-ribosylation factor guanine nucleotide factor 6; AI661273; BC003498; D430018P08; EFA6D; EFA6R; Epididymis tissue protein Li 20mP; exchange factor for ADP-ribosylation factor guanine nucleotide factor 6; Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6 D; Exchange factor for ARF6 D; guanine nucleotide exchange factor for ADP ribosylation factor 6; HCA67; Hepatocellular carcinoma-associated antigen 67; KIAA0942; PH and SEC7 domain-containing protein 3; pleckstrin and Sec7 domain containing 3; pleckstrin homology and SEC7 domain-containing protein 3; Psd3; RGD1559968 | |
Rabbit | |
Protein A | |
RUO | |
23362 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NYI0 | |
PSD3 | |
Synthetic peptide directed towards the middle region of human PSD3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction