Learn More
PSEN2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 161681AP
Catalog No. 50-173-0767
Add to cart
Description
Description
This antibody is specific to presenilin 2. It will not bind presenilin 1.
Human SUG1 (also known as p45, Rpt6 and Cim3) is a component of the 19S regulatory subunit of the 26S proteosome complex. It is one of the AAA ATPas found in the 19S regulatory complex that is thought to participate in the unfolding of ubiquitinated proteins in an ATP-dependent manner. It has also been shown to directly interact with the nucleotide excision repair protein XPB.
Specifications
Specifications
PSEN2
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P49810, Q61144
AD3L, AD3LP, AD4, AD5, E5 1, Presenilin 2, PS 2, PS2, PSEN2, PSNL2, STM 2, STM2
Rabbit
IgG
150 μL
-20°C
Primary
19165, 5664
Immunohistochemistry (Paraffin)
Unconjugated
PSEN2
Liquid
Psen2
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse