Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pseudomonas Aeruginosa Mouse anti-Bacteria, Clone: 1200/472, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | Pseudomonas Aeruginosa |
---|---|
Clone | 1200/472 |
Concentration | 0.2 mg/ml |
Dilution | ELISA 1-5 ug/ml |
Applications | ELISA |
Description
Pseudomonas Aeruginosa Monoclonal specifically detects Pseudomonas Aeruginosa in Bacteria samples. It is validated for ELISA.Specifications
Pseudomonas Aeruginosa | |
0.2 mg/ml | |
ELISA | |
Unconjugated | |
Mouse | |
Bacteria | |
Pseudomonas aeruginosa 9 | |
IgG1 κ | |
Protein A or G purified |
1200/472 | |
ELISA 1-5 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
Pseudomonas aeruginosa serotype 6C | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title