Learn More
PseudomonasAeruginosa Mouse anti-Bacteria, Janelia Fluor 646, Clone: 1001/214, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP125946JF646
Catalog No. NBP12946646
Add to cart
Description
Description
PseudomonasAeruginosa Monoclonal antibody specifically detects PseudomonasAeruginosa in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
PseudomonasAeruginosa
1001/214
Immunocytochemistry/Immunofluorescence
Purified
Mouse
IgG3
0.1mL
Primary
NBP1-25946 is specific for P. aeruginosa serotype 3.
Immunocytochemistry, Immunofluorescence
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Pseudomonas aeruginosa 9
P. aeruginosa serotype 3 (ATCC 33350)
Protein A purified
Store at 4°C in the dark.
Monoclonal
Bacteria