Learn More
PseudomonasAeruginosa Mouse anti-Bacteria, Janelia Fluor 646, Clone: 95/159, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB10064562JF646
Catalog No. NB064562646
Add to cart
Description
Description
PseudomonasAeruginosa Monoclonal antibody specifically detects PseudomonasAeruginosa in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
PseudomonasAeruginosa
95/159
Immunocytochemistry/Immunofluorescence
Purified
Mouse
IgG1
0.1mL
Primary
Specific for Pseudomonas aeruginosa, serotype 9.
Immunocytochemistry, Immunofluorescence
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Pseudomonas aeruginosa 9
Pseudomonas aeruginosa, serotype 9 (NCTC 11448, ATCC 33356).
Protein A purified
Store at 4°C in the dark.
Monoclonal
Bacteria