Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pseudothiohydantoin 97%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Plastic bottle
556-90-1
C3H4N2OS
116.138
MFCD00205270
HYMJHROUVPWYNQ-UHFFFAOYSA-N
pseudothiohydantoin,2-aminothiazol-4 5h-one,2-imino-4-thiazolidinone,4 5h-thiazolone, 2-amino,usaf dm-1,2-imino-4-thiazolidone,2-amino-2-thiazolin-4-one,usaf be-4-5,2-amino-4-oxo-thiazole,2-imino-thiazolidin-4-one
11175
2-amino-1,3-thiazol-4-one
C1C(=O)N=C(S1)N
556-90-1
C3H4N2OS
116.138
MFCD00205270
HYMJHROUVPWYNQ-UHFFFAOYSA-N
pseudothiohydantoin,2-aminothiazol-4 5h-one,2-imino-4-thiazolidinone,4 5h-thiazolone, 2-amino,usaf dm-1,2-imino-4-thiazolidone,2-amino-2-thiazolin-4-one,usaf be-4-5,2-amino-4-oxo-thiazole,2-imino-thiazolidin-4-one
11175
2-amino-1,3-thiazol-4-one
C1C(=O)N=C(S1)N
Quantity 25g
Color White to Yellow
Infrared Spectrum Authentic
Packaging Plastic bottle
Assay Percent Range 97%
Beilstein 27, 233
Formula Weight 116.14
Physical Form Fine Needles or Powder
Percent Purity ≥96.0%
Chemical Name or Material Pseudothiohydantoin, 97%