Learn More
PSG1 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29928
Catalog No. 89-033-216
Add to cart
Description
Description
Rabbit polyclonal antibody raised against synthetic peptide of human PSG1.
  • High specificity
  • High reactivity
Specifications
Specifications
PSG1
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (2.5 ug/mL) The optimal working dilution should be determined by the end user.
NM_006905
B1G1|CD66f|DHFRP2|FLJ90598|FLJ90654|PBG1|PSBG1|PSGGA|PSGIIA|SP1
Rabbit
Protein A purification
RUO
Primary
5669
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of human PSG1.
In PBS (2% sucrose, 0.09% sodium azide).
Liquid
PSG1
A synthetic peptide corresponding to N-terminus of human PSG1.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)