Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PSG11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113510
Description
PSG11 Polyclonal Antibody for Western Blot, ELISA
PSG11, also named as PSG13, PSG14, PSBG-11 and PSBG-13, belongs to the immunoglobulin superfamily and CEA family. PSBG are produced in high quantity during pregnancy. PSG11 and other PSGs induce dose-dependent secretion of anti-inflammatory cytokines by human monocytes.
Specifications
PSG11 | |
Polyclonal | |
Unconjugated | |
PSG11 | |
pregnancy specific beta-1-glycoprotein 11; pregnancy-specific beta-1-glycoprotein 11; Pregnancy-specific beta-1-glycoprotein 13; PS-beta-G-11; PS-beta-G-13; PSBG-11; PSBG-13; PSG11; PSG13; PSG14 | |
Rabbit | |
Protein G | |
RUO | |
5680 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9UQ72 | |
PSG11 | |
Recombinant Human Pregnancy-specific beta-1-glycoprotein 11 protein (7-219AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction