Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSG16 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP267085

Catalog No. NBP267085


Add to cart

Description

Description

PSG16 Polyclonal antibody specifically detects PSG16 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PSG16
Unconjugated
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide
Rabbit
IgG
100μL
Primary
Mouse
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Western Blot 1:1000, Flow Cytometry, Immunocytochemistry/Immunofluorescence 1:100-1:200
Affinity Purified
Synthetic peptide within human Psg16 aa 360-372.
Peptide affinity purified
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit