Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSG2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571976
Description
The human pregcy-specific glycoproteins (PSGs) are a family of proteins that are synthesized in large amounts by placental trophoblasts and released into the maternal circulation during pregcy. Molecular cloning and analysis of several PSG genes has indicated that the PSGs form a subgroup of the carcinoembryonic antigen (CEA) gene family, which belongs to the immunoglobulin superfamily of genes. Members of the CEA family consist of a single N domain, with structural similarity to the immunoglobulin variable domains, followed by a variable number of immunoglobulin constant-like A and/or B domains. Most PSGs have an arg-gly-asp (RGD) motif, which has been shown to function as an adhesion recognition signal for several integrins, in the N-terminal domain (summary by Teglund et al., 1994 [PubMed 7851896]). For additional general information about the PSG gene family, see PSG1 (MIM 176390).
Specifications
PSG2 | |
Polyclonal | |
Unconjugated | |
PSG2 | |
Pregcy-specific beta-1-glycoprotein 2; PS-beta-G-2; PSBG-2; Pregcy-specific glycoprotein 2; Pregcy-specific beta-1 glycoprotein E; PS-beta-E; PSG2; PSBG2 | |
Rabbit | |
Protein A | |
RUO | |
5670 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.06 mg/mL | |
PBS with 0.09% sodium azide | |
P11465 | |
PSG2 | |
KLH conjugated synthetic peptide between 225-254 amino acids from the C-terminal region of human PSG2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction