Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSG9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219979
Description
PSG9 Polyclonal specifically detects PSG9 in Human samples. It is validated for Western Blot.Specifications
PSG9 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
pregnancy specific beta-1-glycoprotein 9, Pregnancy-specific beta-1 glycoprotein B, Pregnancy-specific beta-1-glycoprotein 11, pregnancy-specific beta-1-glycoprotein 9, Pregnancy-specific glycoprotein 11, Pregnancy-specific glycoprotein 7, Pregnancy-specific glycoprotein 9, PS34, PS-beta-B, PS-beta-G-11, PS-beta-G-9, PSBG-11, PSBG-9, PSG11pregnancy specific beta-1-glycoprotein 11, PSG7, PSGII | |
Rabbit | |
48 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot | |
0.65mg/mL | |
Western Blot 1:1000-1:10000 | |
Q00887 | |
PSG9 | |
Recombinant protein encompassing a sequence within the center region of human PSG9. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
5678 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction