Learn More
PSGL-1/CD162 Mouse anti-Human, Alexa Fluor 405, Clone: CHO131, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB996V100UG
Catalog No. FAB996V100
Add to cart
This item is not returnable. View return policy
Description
Description
PSGL-1/CD162 Monoclonal specifically detects PSGL-1/CD162 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
PSGL-1/CD162
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA, SELPLG
Mouse
IgM
RUO
Primary
6404
Human
Flow Cytometry
CHO131
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
SELPLG
CHO Chinese hamster ovary cell line transfected with human PSGL-1/CD162
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human PSGL-1/CD162. Recognizes sLe x -bearing core 2 O-glycan stuctures. It does not recognize sLe x on an extended core 1 O-glycan. The sLe x -bearing, core 2 O-glycan structure decorates the P-Selectin ligand PSGL-1, and the presence of this glycan structure is required for high affinity P-Selectin binding (1). This antibody stains human and canine leukocytes but does not recognize monkey, mouse, rabbit, porcine, feline or bovine leukocytes.
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)