Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSGL-1/CD162 Mouse anti-Human, Alexa Fluor™ 750, Clone: 688124, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9962S100UG
This item is not returnable.
View return policy
Description
PSGL-1/CD162 Monoclonal specifically detects PSGL-1/CD162 in Human samples. It is validated for Flow Cytometry.Specifications
PSGL-1/CD162 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA, SELPLG | |
Chinese hamster ovary cell line CHO-derived recombinant human PSGL-1/CD162, Gln42-Gly295, Accession # Q14242 | |
100 μg | |
6404 | |
Human | |
Supernatant |
Flow Cytometry | |
688124 | |
Flow Cytometry 0.25-1 μg/106 cells | |
Q14242 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human PSGL-1/CD162 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title