Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSGL-1/CD162 Mouse anti-Human, Clone: PSGL1/1567, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen PSGL-1/CD162
Clone PSGL1/1567
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Applications Flow Cytometry, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123114
View Documents
Novus Biologicals
NBP30782020UG
20 μg
Each for $138.37
Add to cart
 
NB123113
View Documents
Novus Biologicals
NBP30782010UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

PSGL-1/CD162 Monoclonal antibody specifically detects PSGL-1/CD162 in Human samples. It is validated for Flow Cytometry, Immunofluorescence
Specifications

Specifications

PSGL-1/CD162
0.2 mg/ml
Flow Cytometry, Immunofluorescence
Unconjugated
RUO
Human
CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA
Recombinant human PSGL-1/CD162 protein
Monoclonal
Column Chromatography
PSGL1/1567
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Unconjugated
Mouse
Cholesterol Metabolism, Immunology, Lipid and Metabolism
10 mM PBS with 0.05% BSA
6404
IgM Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit