Learn More
MBL International Corporation PSMA PE 50 TESTSPSMA PE 50 DFS Item
Manufacturer:  MBL International Corporation K01425
PSMA Monoclonal Antibody, PE Labeled Size 50 tests Clone Number 107-1A4 Host Species Mouse; Conjugate PE Application FCM
Catalog No. 50-481-84