Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMA4 Mouse anti-Human, Clone: OTI2D12, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525838

Catalog No. PIMA525838


Add to cart

Description

Description

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. Three alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

PSMA4
Monoclonal
0.79 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P25789
HC9;HsT17706;Macropain subunit C9;MGC111191;MGC12467;MGC24813;Multicatalytic endopeptidase complex subunit C9;Proteasome (prosome macropain) subunit alpha type 4;Proteasome alpha 4 subunit;Proteasome component C9;Proteasome subunit alpha type 4;Proteasome subunit alpha type-4;Proteasome subunit HC9;Proteasome subunit L;PSA4_HUMAN;PSC9;PSMA 4;psmA4;
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5685
Flow Cytometry, Western Blot
OTI2D12
Unconjugated
PSMA4
Liquid
PSMA4
Full length human recombit protein of PSMA4 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit