Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMA6 Monoclonal Antibody (OTI4C9), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA526371
Description
PSMA6 Monoclonal antibody specifically detects PSMA6 in Canine, Human, Mouse, Non-human Primate, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
PSMA6 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P60900, P60901, Q9QUM9 | |
PSMA6 | |
Full length human recombinant protein of PSMA6 produced in E.coli | |
100 μL | |
Primary | |
Canine, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG2b |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI4C9 | |
Unconjugated | |
PSMA6 | |
27 kDa prosomal protein; EGK_18113; IOTA; Macropain iota chain; macropain subunit iota; MGC22756; MGC2333; MGC23846; multicatalytic endopeptidase complex iota chain; p27K; PROS 27; PROS27; PROS-27; prosomal P27K protein; proteasome (prosome, macropain) subunit, alpha type 6; proteasome (prosome, macropain) subunit, alpha type, 6; proteasome 20S subunit alpha 6; proteasome alpha 6 subunit; proteasome iota chain; proteasome subunit alpha 6; proteasome subunit alpha type-6; proteasome subunit iota; PSA6; PSMA 6; PSMA6; testicular secretory protein Li 44 | |
Mouse | |
Affinity Chromatography | |
RUO | |
26443, 29673, 480290, 5687 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction