Learn More
PSMA7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 152191AP
Catalog No. 50-173-0075
Add to cart
Description
Description
Component of the 20S core proteasome complex involved in the proteolytic degradation of most intracellular proteins. This complex plays numerous essential roles within the cell by associating with different regulatory particles. Associated with two 19S regulatory particles, forms the 26S proteasome and thus participates in the ATP-dependent degradation of ubiquitinated proteins. The 26S proteasome plays a key role in the maintece of protein homeostasis by removing misfolded or damaged proteins that could impair cellular functions, and by removing proteins whose functions are no longer required. Associated with the PA200 or PA28, the 20S proteasome mediates ubiquitin-independent protein degradation. This type of proteolysis is required in several pathways including spermatogenesis (20S-PA200 complex) or generation of a subset of MHC class I-presented antigenic peptides (20S-PA28 complex). Inhibits the transactivation function of HIF-1A under both normoxic and hypoxia-mimicking conditions. The interaction with EMAP2 increases the proteasome-mediated HIF-1A degradation under the hypoxic conditions. Plays a role in hepatitis C virus internal ribosome entry site-mediated translation. Mediates nuclear translocation of the androgen receptor (AR) and thereby enhances androgen-mediated transactivation.
Specifications
Specifications
PSMA7
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O14818, P48004, Q9Z2U0
20S proteasome, C6, HSPC, Proteasome subunit RC6 1, Proteasome subunit XAPC7, PSMA7, RC6 1, XAPC7
Rabbit
IgG
150 μL
-20°C
Primary
26444, 29674, 5688
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
PSMA7
Liquid
PSMA7
PSMA7 Fusion Protein Ag7389
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat