Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMB1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549648

Catalog No. PA549648


Add to cart

Description

Description

The proteasome is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. The proteasome has an ATP-dependent proteolytic activity.
Specifications

Specifications

PSMB1
Polyclonal
PBS with 0.09% sodium azide
P20618, O09061
Proteasome subunit beta type-1, Macropain subunit C5, Multicatalytic endopeptidase complex subunit C5, Proteasome component C5, Proteasome gamma chain, PSMB1, PSC5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
19170, 5689
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PSMB1
Liquid
PSMB1
KLH conjugated synthetic peptide between 214-241 amino acids from the C-terminal region of human PSMB1.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit