Learn More
PSMB5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 191781AP
Catalog No. 50-173-2360
Add to cart
Description
Description
Cyclic nucleotide phosphodiesterases.
Specifications
Specifications
PSMB5
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O55234, P28074, P28075
20S proteasome, LMPX, Macropain epsilon chain, MB1, Proteasome chain 6, Proteasome epsilon chain, Proteasome subunit beta type 5, Proteasome subunit MB1, Proteasome subunit X, PSMB5, X
Rabbit
IgG
150 μL
-20°C
Primary
19173, 29425, 5693
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PSMB5
Liquid
Psmb5
PSMB5 Fusion Protein Ag5828
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat