Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PSMC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | PSMC1 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119362
|
Invitrogen™
PA5119362 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121665
|
Invitrogen™
PA5121665 |
20 μL |
Each for $209.50
|
|
|||||
Description
PSMC1 Polyclonal Antibody for Western Blot, IHC (P)
The 26S protease is involved in the ATP-dependent degradation of ubiquitinated proteins. The regulatory (or ATPase) complex confers ATP dependency and substrate specificity to the 26S complex.Specifications
PSMC1 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
Psmc1 | |
26S protease regulatory subunit 4; 26S proteasome AAA-ATPase subunit RPT2; 26S proteasome regulatory subunit 4; AI325227; MGC24583; MGC8541; P26s4; p56; peptidase (prosome, macropain) 26S subunit, ATPase 1; protease (prosome, macropain) 26S subunit, ATPase 1; proteasome (prosome, macropain) 26S subunit, ATPase, 1; proteasome 26S ATPase subunit 1; proteasome 26S subunit ATPase 1; proteasome 26S subunit, ATPase 1; proteasome 26S subunit, ATPase, 1; prs4; Psmc1; rpt2; Rpt2/S4; S4 | |
Psmc1 | |
Primary | |
Antigen Affinity Chromatography |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P62191 | |
5700 | |
Recombinant protein Proteasome 26S Subunit, ATPase 1. The antigen corresponds to amino acid range 1-440 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title