Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMC3 Monoclonal Antibody (OTI9G5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA500929S


Description
PSMC3 Monoclonal antibody specifically detects PSMC3 in Canine, Human, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
PSMC3 | |
Monoclonal | |
0.94 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P17980, Q63569 | |
Mouse | |
Affinity chromatography | |
RUO | |
29677, 475980, 5702 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunocytochemistry, Western Blot | |
OTI9G5 | |
Unconjugated | |
PSMC3 | |
26S protease regulatory subunit 6A; 26S proteasome AAA-ATPase subunit RPT5; 26S proteasome regulatory subunit 6A; human immunodeficiency virus tat transactivator binding protein-1; MGC8487; proteasome (prosome, macropain) 26S subunit, ATPase 3; proteasome (prosome, macropain) 26S subunit, ATPase, 3; proteasome 26S subunit ATPase 3; proteasome 26S subunit, ATPase 3; proteasome subunit P50; Psmc3; rpt5; Spermatogenic cell/sperm-associated Tat-binding protein homolog SATA; Tat binding protein 1; Tat-binding protein 1; TBP1; TBP-1; testicular secretory protein Li 42 | |
Full length human recombinant protein of human PSMC3 produced in HEK293T cell. | |
30 μL | |
Primary | |
Canine, Human, Rat | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction