Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMD11 Mouse, DyLight 680, Clone: 1F4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP130252FR

Catalog No. NBP130252FR


Add to cart

Description

Description

PSMD11 Monoclonal antibody specifically detects Antigen in C. elegans, Human, Mouse samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications

Specifications

PSMD11
1F4
Western Blot, ELISA
Purified
PSMD11
Recombinant human PSMD11 (1-422aa) purified from E. coli (NP_002806).
Protein A or G purified
RUO
Primary
5717
C. elegans, Human, Mouse
ELISA, Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
MGC3844,26S proteasome regulatory subunit RPN6, p44.5,26S proteasome regulatory subunit 9, proteasome (prosome, macropain) 26S subunit, non-ATPase, 11,26S proteasome regulatory subunit p44.5, Rpn6,26S proteasome regulatory subunit S9, S9,26S proteasome non-ATPase regulatory subunit 11
Mouse
IgG1 Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PSMD11 protein.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit