Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMD13 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549551

Catalog No. PA549551

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse and rat based on sequence homology.

Acts as a regulatory subunit of the 26S proteasome which is involved in the ATP-dependent degradation of ubiquitinated proteins.
Specifications

Specifications

PSMD13
Polyclonal
PBS with 0.09% sodium azide
Q9UNM6
26S proteasome non-ATPase regulatory subunit 13, 26S proteasome regulatory subunit RPN9, 26S proteasome regulatory subunit S11, 26S proteasome regulatory subunit p405, PSMD13
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5719
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PSMD13
Liquid
PSMD13
KLH conjugated synthetic peptide between 269-298 amino acids from the C-terminal region of human PSMD13.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit