Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PSMD2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518324
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes one of the non-ATPase subunits of the 19S regulator lid. In addition to participation in proteasome function, this subunit may also participate in the TNF signalling pathway since it interacts with the tumor necrosis factor type 1 receptor. A pseudogene has been identified on chromosome 1.
Specifications
PSMD2 | |
Polyclonal | |
Unconjugated | |
PSMD2 | |
26S proteasome non-ATPase regulatory subunit 2; 26S proteasome non-ATPase regulatory subunit 2-like protein; 26S proteasome regulatory subunit RPN1; 26S proteasome regulatory subunit S2; 26S proteasome subunit p97; 55.11 protein; 9430095H01Rik; AA407121; F23F1.8; I79_017809; MGC14274; P97; proteasome (prosome, macropain) 26S subunit, non-ATPase, 2; proteasome 26S subunit, non-ATPase 2; protein 55.11; PSMD2; RPN1; S2; TEG-190; testis expressed gene 190; Tex190; TNFR-associated protein 2; TRAP2; Tumor necrosis factor type 1 receptor-associated protein 2 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
21762, 287984, 5708 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q13200, Q4FZT9, Q8VDM4 | |
PSMD2 | |
Synthetic peptide sequence (VILRKNPNYDL) corresponding to the C-terminus amino acids of PSMD2 (aa 898-908). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction