Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMD3 Rabbit anti-Human, Mouse, Alexa Fluor® 647, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1974A647

Catalog No. PIPA1974AY


Add to cart

Description

Description

PA1974A647 detects proteasome 19S subunit S3 from human and mouse cells. PA1974A647 immunizing peptide corresponds to amino acid residues 513-534 from human proteasome 19S subunit S3 protein.

Proteolytic degradation is critical to the maintece of appropriate levels of short-lived and regulatory proteins as important and diverse as those involved in cellular metabolism, heat shock and stress response, antigen presentation, modulation of cell surface receptors and ion channels, cell cycle regulation, transcription, and signalling factors. The ubiquitin-proteasome pathway deconstructs most proteins in the eukaryotic cell cytosol and nucleus. Others are degraded via the vacuolar pathway which includes endosomes, lysosomes, and the endoplasmic reticulum. The 26S proteasome is an ATP-dependent, multisubunit (~31), barrel-shaped molecular machine with an apparent molecular weight of ~2.5 MDa. It consists of a 20S proteolytic core complex which is crowned at one or both ends by 19S regulatory subunit complexes. The 19S regulatory subunits recognize ubiquitinated proteins and play an essential role in unfolding and translocating targets into the lumen of the 20S subunit. An enzymatic cascade is responsible for the attachment of multiple ubiquitin molecules to lysine residues of proteins targeted for degradation. Several genetic diseases are associated with defects in the ubiquitin-proteasome pathway. Some examples of affected proteins include those linked to cystic fibrosis, Angelman""e;s syndrome, and Liddle syndrome.
Specifications

Specifications

PSMD3
Polyclonal
Alexa Fluor™ 647
PSMD3
Liquid
PSMD3
Synthetic Peptide: E(513) R E Q Q D L E F A K E M A E D D D D S F P(534)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunofluorescence
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
O43242, P14685
AI255837, S3, D11Bwg1349e, Tstap91a, TSTA2, RPN3, Psd3, P58, AntP91a
Rabbit
IgG
50 μL
4° C, store in dark, DO NOT FREEZE!
Primary
22123, 5709
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only