Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMD9 Mouse anti-Human, Clone: OTI5H2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527265

Catalog No. PIMA527265


Add to cart

Description

Description

The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a non-ATPase subunit of the 19S regulator.
Specifications

Specifications

PSMD9
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O00233
26S proteasome non-ATPase regulatory subunit 9; 26S proteasome regulatory subunit p27; Homolog of rat Bridge 1; p27; Proteasome (prosome macropain) 26S subunit non ATPase 9; Proteasome 26S non ATPase regulatory subunit 9; Proteasome 26S subunit non ATPase 9; Psmd9; PSMD9_HUMAN; Rpn4;
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5715
Western Blot
OTI5H2
Unconjugated
PSMD9
Liquid
PSMD9
Full length human recombit protein of PSMD9 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit