Learn More
Aviva Systems Biology Corp PSME1 Antibody - middle region (ARP58519_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58519_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSME1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PSME1. Alternate names: IFI5111, MGC8628, PA28A, PA28alpha, REGalphaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-62338