Learn More
PSME4 Rabbit anti-Human, Alexa Fluor™ 750, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222236AF750
Catalog No. NP222236A70
Add to cart
Description
Description
PSME4 Polyclonal antibody specifically detects PSME4 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
PSME4
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
PSME4
The immunogen this antibody was made to, maps to a region between residue 1 and 50 of human Proteasome (Prosome, Macropain) Activator subunit 4 using the numbering given in entry NP_055429.2 (GeneID 23198).
100 μL
Primary
23198
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
proteasome (prosome, macropain) activator subunit 4
Rabbit
IgG
Store at 4°C in the dark.
Polyclonal
Human