Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMG2 Mouse anti-Human, Clone: OTI1E8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526947

Catalog No. PIMA526947


Add to cart

Description

Description

Chaperone protein which promotes assembly of the 20S proteasome as part of a heterodimer with PSMG1. The PSMG1-PSMG2 heterodimer binds to the PSMA5 and PSMA7 proteasome subunits, promotes assembly of the proteasome alpha subunits into the heteroheptameric alpha ring and prevents alpha ring dimerization.
Specifications

Specifications

PSMG2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q969U7
CLAST3, HCCA3, HsT1707, MDS003, PAC2, TNFSF5IP1, CD40 ligand-activated specific transcript 3, HDCMC29P, HSPC260, PAC-2, hepatocellular carcinoma susceptibility protein, hepatocellular carcinoma-susceptibility protein 3, likely ortholog of mouse CD40 ligand-activated specific transcript 3 (Clast3), proteasome a
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56984
Immunohistochemistry (Paraffin), Western Blot
OTI1E8
Unconjugated
PSMG2
Liquid
PSMG2
Full length human recombit protein of PSMG2 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit