Learn More
PSMG3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 156011AP
Catalog No. 50-173-0340
Add to cart
Description
Description
PSMG3, also named as C7orf48 and PAC3, is a chaperone protein which promotes assembly of the 20S proteasome. It may cooperate with PSMG1-PSMG2 heterodimers to orchestrate the correct assembly of proteasomes.
Specifications
Specifications
PSMG3
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BT73, Q9CZH3
C7orf48, hPAC3, PAC 3, PAC3, PSMG3
Rabbit
IgG
150 μL
-20°C
Primary
100361517, 66506, 84262
Immunohistochemistry (Paraffin)
Unconjugated
PSMG3
Liquid
Psmg3
PSMG3 Fusion Protein Ag7984
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat