Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ PSNCBAM-1
CB1 receptor negative allosteric modulator
$299.28 - $1248.16
Specifications
Purity | >99% |
---|---|
Product Type | PSNCBAM-1 |
Molecular Weight (g/mol) | 392.88 |
Inhibitors | PSNCBAM-1 |
Description
CB1 receptor negative allosteric modulator (IC50 values are 45 and 209 nM for the inhibition of CP 55,940 and WIN 55,212-2 respectively). Has no effect at the CB2 receptor. Decreases food intake and body weight in rats.Specifications
>99% | |
PSNCBAM-1 | |
392.88 | |
PSNCBAM-1 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title