Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSP/BPIFA2 Mouse anti-Human, Biotin, Clone: 3F8E4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237277B

Catalog No. NBP237277B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

PSP/BPIFA2
3F8E4
Western Blot, ELISA, Immunohistochemistry
Purified
BPIFA2
Purified recombinant fragment of mouse mSplunc2 (AA: 16-169) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot, ELISA, Immunohistochemistry
Biotin
PBS with 0.05% Sodium Azide
bA49G10.1, chromosome 20 open reading frame 70, Parotid secretory protein, PSP, short palate, lung and nasal epithelium carcinoma associated 2, short palate, lung and nasal epithelium carcinoma-associated protein 2, SPLUNC2
Mouse
IgG1
0.1 ml
Primary
140683
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit