Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSP94/MSMB Rabbit anti-Human, Mouse, Clone: 4L8B5, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | PSP94/MSMB |
---|---|
Clone | 4L8B5 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PSP94/MSMB Monoclonal antibody specifically detects PSP94/MSMB in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
PSP94/MSMB | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
4477 | |
IgG | |
Affinity purified |
4L8B5 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
IGBF, MSMB, MSPB, PIP, PN44, PRPS, PSP57, PSP94 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PSP94/MSMB (P08118). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title