Learn More
Aviva Systems Biology Corp PSTK Antibody - middle region (ARP53154_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53154_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSTK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PSTK. Alternate names: C10orf89, MGC35392ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-60946