Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-PSTPIP1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP185764
Description
PSTPIP1 Polyclonal antibody specifically detects PSTPIP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PSTPIP1 | |
Unconjugated | |
Affinity Purified | |
CD2BP1CD2-binding protein 1, CD2BP1L, CD2BP1S, H-PIPPEST phosphatase-interacting protein 1, PAPASCD2 antigen-binding protein 1, proline-serine-threonine phosphatase interacting protein 1, proline-serine-threonine phosphatase-interacting protein 1, PSTPIPCD2 cytoplasmic tail-binding protein | |
PSTPIP1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GMIKRFSGLLHGSPKTTSLAASAASTETLTPTPERNEGVYTAIAVQEIQGNPASPAQEYRALYDYTAQNPDELDLSAGDILEVILEGEDGWWTVERNGQRGFVPGSYLEK | |
Polyclonal | |
Immunogen affinity purified | |
RUO | |
Human |
Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin) | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
9051 | |
Rabbit | |
IgG | |
Primary | |
0.1 ml | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only