Learn More
Metrohm USA PT TITRODE
Supplier: Metrohm USA 60431100
Double Pt-wire electrode for redox titrations without change of pH value Maintenance-free combined Pt-ring electrode with pH membrane as reference electrode