Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTAC oxalate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Muscarinic receptor ligand. Displays partial agonist activity at M2 and M4 receptors; exhibits antagonist effects at M1, M3 and M5 receptors (Ki values are 2.8, 0.2, 0.6, 0.2 and 0.8 nM respectively). Displays selectivity for muscarinic receptors over a range of neurotransmitter receptors and ion channels. Inhibits firing rate of dopaminergic cells in the limbic ventral tegmental area after acute administration, without binding to dopamine receptors.
Specifications
Quantity | 10 mg |
Formula Weight | 359.46 |
Percent Purity | >98% |
Chemical Name or Material | PTAC oxalate |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction