Learn More
PTCH1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 175201AP
Catalog No. 50-173-1622
Add to cart
Description
Description
This gene encodes a member of the patched gene family. The encoded protein is the receptor for sonic hedgehog, a secreted molecule implicated in the formation of embryonic structures and in tumorigenesis, as well as the desert hedgehog and indian hedgehog proteins. This gene functions as a tumor suppressor. Mutations of this gene have been associated with basal cell nevus syndrome, esophageal squamous cell carcinoma, trichoepitheliomas, transitional cell carcinomas of the bladder, as well as holoprosencephaly. Alternative splicing results in multiple transcript variants encoding different isoforms. Additional splice variants have been described, but their full length sequences and biological validity cannot be determined currently.
Specifications
Specifications
PTCH1
0.48 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13635, Q61115
HPE7, NBCCS, Patched, patched homolog 1 (Drosophila), Patched1, Protein patched homolog 1, PTC, PTC1, PTCH, PTCH1, PTCH11
Rabbit
IgG
150 μL
-20°C
Primary
19206, 5727
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PTCH1
Liquid
PTCH1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse