Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTCHD3 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA538002
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
May play a role in sperm development or sperm function.
Specifications
PTCHD3 | |
Polyclonal | |
Unconjugated | |
PTCHD3 | |
4930451E13Rik; 4933440L20Rik; FLJ44037; MGC129888; patched domain containing 3; patched domain-containing protein 3; Patched-related protein; PTCHD3; PTR; RNDEu-3; RND-type protein RNDEu-3 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
374308 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q3KNS1 | |
PTCHD3 | |
Peptide with sequence C-KKVDYWDKDVRQ, from the internal region of PTCHD3 (aa 663-675). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction