Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTCRA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110236
Description
Immunogen sequence: LCDPAGPLPS PATTTRLRAL GSHRLHPATE TGGREATSSP RPQPRDRRWG DTPPGRKPGS PVWGEGSYLS SYPTCPAQAW CSRSALRAPS SSLGAFFAGD LPPPLQAGAA.
The pre-T-cell receptor complex (composed of PTCRA, TCRB and the CD3 complex) regulates early T-cell development.
Specifications
PTCRA | |
Polyclonal | |
Unconjugated | |
PTCRA | |
pre T-cell antigen receptor alpha; pre-T-cell receptor alpha chain; PTA; pT-alpha; pT-alpha-TCR; PTCRA | |
Rabbit | |
Affinity chromatography | |
RUO | |
171558 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.57 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6ISU1 | |
PTCRA | |
Recombinant fusion protein containing a sequence corresponding to amino acids 172-281 of human PTCRA (NP_612153.2). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction