Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTEN-Long Mouse anti-Human, Clone: 1C2, MilliporeSigma™
Mouse Monoclonal Antibody
$177.00 - $450.00
Specifications
Antigen | PTEN-Long |
---|---|
Clone | 1C2 |
Concentration | 1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABS450B25
|
MilliporeSigma
MABS450B25UG |
25 μg |
Each for $177.00
|
|
|||||
MABS450B100
|
MilliporeSigma
MABS450B100UG |
100 μg |
Each for $450.00
|
|
|||||
Description
PTEN-Long Monoclonal antibody specifically detects PTEN-Long in Human samples. It is validated for ELISA, Western Blotting.Specifications
PTEN-Long | |
1.0 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
1C2 | |
ELISA, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
A bacterial fusion protein with a V5/His-tag at its C- terminus. | |
Protein G purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title