Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTEN/PTEN-Long Mouse anti-Human, Clone: 1A1, MilliporeSigma™

Mouse Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen PTEN/PTEN-Long
Clone 1A1
Concentration 1.0 mg/mL
Applications ELISA
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABC1722100
View Documents
MilliporeSigma
MABC1722100UG
100 μg
Each for $373.00
Add to cart
 
MABC172225
View Documents
MilliporeSigma
MABC172225UG
25 μg
Each for $145.80
Add to cart
 
Description

Description

PTEN/PTEN-Long Monoclonal antibody specifically detects PTEN/PTEN-Long in Human samples. It is validated for ELISA.
Specifications

Specifications

PTEN/PTEN-Long
1.0 mg/mL
Unconjugated
Mouse
Human
A bacterial fusion protein with a V5/His-tag at its C- terminus.
Monoclonal
Protein G purified
1A1
ELISA
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG1κ
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit