Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTEN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541332
Description
Peptide sequence: KILIDVFDGP PLYDDVKVQF FYSNLPTYYD NCSFYFWLHT SFIENNRLYL Sequence homology: Human: 100%.
TPTE encodes a putative transmembrane tyrosine phosphatase that may be involved in signal transduction pathways of the endocrine or spermatogenetic function of the testis.
Specifications
PTEN2 | |
Polyclonal | |
Unconjugated | |
TPTE | |
Cancer/testis antigen 44; CT44; Pten2; PTEN-related tyrosine phosphatase; Putative tyrosine-protein phosphatase TPTE; tensin, putative protein-tyrosine phosphatase; TPTE; Transmembrane phosphatase with tensin homology; tumor antigen BJ-HCC-5 | |
Rabbit | |
Protein A | |
RUO | |
7179 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P56180 | |
TPTE | |
Synthetic peptide directed towards the C-terminal of human TPTE. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction