Learn More
Aviva Systems Biology Corp PTF1A ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP32531_P050
This is a rabbit polyclonal antibody against PTF1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PTF1A. Alternate names: PTF1-p48, bHLHa29ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat, Yeast
Catalog No. 50-014-54637