Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTF1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112677
Description
PTF1A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
PFT1A is a component of the pancreas transcription factor 1 complex and is known to have a role in mammalian pancreatic development. The protein plays a role in determining whether cells allocated to the pancreatic buds continue towards pancreatic organogenesis or revert back to duodenal fates. The protein is thought to be involved in the maintenance of exocrine pancreas-specific gene expression including elastase 1 and amylase. Mutations in this gene cause cerebellar agenesis and loss of expression is seen in ductal type pancreas cancers.
Specifications
PTF1A | |
Polyclonal | |
Unconjugated | |
PTF1A | |
bHLH transcription factor; bHLH transcription factor p48; BHLHA29; class A basic helix-loop-helix protein 29; class II bHLH protein PTF1A; exocrine pancreas-specific transcription factor p48; p48 DNA-binding subunit of transcription factor PTF1; PACA; PAGEN2; pancreas associated transcription factor 1a; pancreas specific transcription factor, 1a; pancreas transcription factor 1 subunit alpha; pancreas transcription factor1 p48 subunit; pancreas-specific transcription factor 1a; PTF1; PTF1A; PTF1p48; PTF1-p48; Ptfa; si:zc142h2.2; zgc:112216 | |
Rabbit | |
Protein G | |
RUO | |
117034, 19213, 256297 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q64305, Q7RTS3, Q9QX98 | |
PTF1A | |
Recombinant Human Pancreas transcription factor 1 subunit alpha protein (224-325AA). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction