Learn More
Chromatography Research Supplies IncSupplier Diversity Partner PTFE FERRULE SET 1/16INOD 10PK DFS Item
Manufacturer:  Chromatography Research Supplies IncSupplier Diversity Partner 214100
100% PTFE; temperature limit 250 deg. C. 1/16 in. straight ferrule fits over 1/16 in. OD tubing; fits into standard 1/16 in. Swagelok (registered) / Parker (registered) type fittings. This design requires no back ferrule. Packaged: 3 in. x 3 in. x 1/4 in. Plastic Dial Pack; Weight: 8.5 g
Catalog No. NC0023193