Learn More
Thermo Scientific™ PTFE-Lined Closed Caps
PTFE-Lined Closed Caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials.
Manufacturer:  Thermo Scientific™ B1675800BL
 View more versions of this product
Catalog No. 14-966-104
  Check Availability
Add to cart
Specifications
Description
Thermo Scientific™ PTFE-Lined Closed Caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials. The same high quality PTFE-lined caps used on our Thermo Scientific glass environmental sample containers. The PTFE liner is bonded to the cap so it won't fall out or become separated from the cap.
  • White polypropylene closed top cap with bonded PTFE liner
  • 24-414 cap has a PTFE-faced silicone liner for added cushion and an extra secure seal when used with bottles and VOA vials
Includes:
Cap with bonded liner
Compatible with:
For use with Thermo Scientific glass environmental sample bottles, jars, jugs and vials.
Description & Specifications
Specifications
Manufactured under an ISO certified quality management system.
Polypropylene
24
White
Unprocessed
None
Closed top cap
Closure, with Bonded Liner
58-400mm
PTFE