Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PTFE-Lined Closed Caps

Click to view available options
Level:
Unprocessed
No. per Case:
24
144

Use these caps as replacements for our glass environmental sample bottles, jars, jugs and vials.

Thermo Scientific™ PTFE-Lined Closed Caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials. The same high quality PTFE-lined caps used on our Thermo Scientific glass environmental sample containers. The PTFE liner is bonded to the cap so it won't fall out or become separated from the cap.

  • White polypropylene closed top cap with bonded PTFE liner
  • 24-414 cap has a PTFE-faced silicone liner for added cushion and an extra secure seal when used with bottles and VOA vials

Includes:

Cap with bonded liner

Compatible with:

For use with Thermo Scientific glass environmental sample bottles, jars, jugs and vials.

Closure Included Yes
For Use With (Equipment) Thermo Scientific Glass Environmental Sample Bottles, Jars, Jugs and Vials
Closure Liner PTFE
Shape Round
Certifications/Compliance Manufactured under an ISO certified quality management system.
Closure Size 45-400 mm
Closure Material Polypropylene
Certification Parameters None
Color White
Description Closed top cap
Material PTFE Liner
No. per Case 24
Type Closure with Bonded Liner, Solid Top
Closure Color White
Level Unprocessed
Liner Material PTFE
Show More Show Less